Masterflex® L/S® Precision Pump Tubing, C-Flex® ULTRA, L/S 14; 25 ft
Supplier: Avantor Fluid Handling
Longest life of any C-Flex® formulation.
- Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Single-use custom lengths and configurations available to minimize downtime
- Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
- Tubing Size: L/S 14
<B>Applications</B>: General-purpose lab uses requiring high performance and long life<br /><B>Characteristics</B>: Exceptional tensile and tear strength, good biocompatibility, low spallation, reduced residue and low gas permeability, smooth surface, translucent