Masterflex® Transfer Tubing, Kynar Flex™ 2750 PVDF, 3/16" ID x 1/4" OD; 50 ft
Supplier: Avantor Fluid Handling
Excellent chemical resistance and UV stable.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Single-use custom lengths and configurations available to minimise downtime
- Tubing size: 3/16" ID x 1/4" OD
<B>Applications:</B> High purity and temperature, UV sensitive, or aggressive chemical fluid transfer.<br /><B>Characteristics:</B> Excellent chemical resistance, low TOC, rigid tubing works well with compression and over-molded fittings and adapters. Translucent. <br /><B>Certification:</B> FDA complaint (21 CFR 177.2600), meets USP Class VI requirements, and is made in accordance to cGMP.<br /><B>Temperature range:</B> -40 to 275 °F (-40 to 135 °C)<br /><B>Sterilize:</B> Gamma irradiation, or ethylene oxide.