Masterflex® L/S® Precision Pump Tubing, FDA-Compliant Viton®, L/S 16; 25 ft
Supplier: Avantor Fluid Handling
Good for food and lab applications.
- Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Single-use custom lengths and configurations available to minimise downtime
- Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
- Tubing Size: L/S 16
<B>Applications</B>: Food or laboratory applications where FDA compliance is required, acid and solvent transfer, high-temperature work<br /><B>Characteristics</B>: Good chemical resistance, opaque black.