Masterflex® Ismatec® Pump Tubing, C-Flex®, 0.25 mm ID; 50 ft
Supplier: Avantor Fluid Handling
Meets the demands of various industries.
- Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Tubing Inner Diameter: 0.25 mm
Meets USP Class VI requirements<br />Autoclavable<br />Temp range: -40 to 135 °C (-40 to 275 °F)