Masterflex® Ismatec® Pump Tubing, C-Flex®, 1.42 mm ID; 50 ft
Supplier: Avantor Fluid Handling
VWR Item#MFLX95718-34
Your Organization's Price:
Retrieving
Meets the demands of various industries.
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Tubing Inner Diameter: 1.42 mm
Meets USP Class VI requirements<br />Autoclavable<br />Temp range: -40 to 135 °C (-40 to 275 °F)