Masterflex® L/S® Precision Pump Tubing, C-Flex®, L/S 16; 25 ft
Supplier: Avantor Fluid Handling
VWR Item#MFLX06424-16
Your Organization's Price:
Retrieving
Combines biocompatibility of silicone with chemical resistance similar to Tygon®.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Very low protein binding
  • Heat sealable, weldable
  • Economical tubing
  • Tubing Size: L/S 16
<B>Applications</B>: General-purpose lab uses requiring high performance and long life<br /><B>Characteristics</B>: Exceptional tensile and tear strength, good biocompatibility, low gas permeability, smooth surface, opaque white
See 96123-16 for the irradiated version of this tubing.