Masterflex® L/S® Spooled Precision Pump Tubing, C-Flex®, L/S 17; 400 ft
Supplier: Avantor Fluid Handling
VWR Item#MFLX06427-17
Your Organization's Price:
Retrieving
Combines biocompatibility of silicone with chemical resistance similar to Tygon®.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Very low protein binding
  • Heat sealable, weldable
  • Economical tubing
  • Tubing Size: L/S 17
<B>Applications</B>: General-purpose lab uses requiring high performance and long life.<br /><B>Characteristics</B>: Exceptional tensile and tear strength, good biocompatibility, low gas permeability, smooth surface, opaque white<br />Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.