Masterflex® Flowmeter Acrylic Tripod Base for 03218-50 and 03218-52
Supplier: Avantor Fluid Handling
VWR Item#MFLX03218-58
Your Organization's Price:
Retrieving
Easily mount flowmeters on tripod for easy viewing.
  • Constructed of durable acrylic
  • Suitable for use with 32461-Series flowmeters
These tripod bases are ideal for benchtop applications. A tripod base is required to use the multitube frame with the common inlet flow pattern; all other multitube frames are designed for panel and tripod mounting. Tripod bases are made of acrylic and include a spirit leveler.