Masterflex® L/S® Small Cartridge for Multichannel Cartridge Pump Head, Microbore and Precision Tubing
Supplier: Avantor Fluid Handling
VWR Item#MFLX07519-80
Your Organization's Price:
Retrieving
Use with L/S® multichannel cartridge pump heads for synchronous, multichannel pumping.
  • Only Silicone and C-Flex® tubing formulations are recommended for size L/S® 17 tubing when used in L/S® cartridge pump heads.
  • Suitable for use with multichannel pump heads 07519-05 and 07519-06
Cartridges feature accurate flow rates and adjustable tubing retainers to hold tubing in place — for large cartridges, set tubing retainers to your tube size and you are ready to pump. On small cartridges, optimum retention is preset — simply load tubing and start pumping. <br /><br />Cartridges are constructed from polycarbonate and glass-filled nylon. Mix small and large cartridges on pump heads that accept both sizes; two small cartridges occupy the same space as one large cartridge.