Masterflex® Fitting, Nylon, Straight, Male Luer Lock to Hose Barb Adapter, 3/32" ID; 25/PK
Supplier: Avantor Fluid Handling
VWR Item#MFLX45505-02
Your Organization's Price:
Retrieving
Ensure your process integrity with full material traceability.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: Male Luer Lock x 3/32" ID
  • Ideal for use with flexible tubing
Hose barbs are available from ¹/₁₆" to ¹/₄" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.