Masterflex® L/S® Tube Set, PTFE, 4 mm ID x 6 mm OD; Set of Two
Supplier: Avantor Fluid Handling
VWR Item#MFLX77390-60
Your Organization's Price:
Retrieving
Excellent chemical resistance and chemically inert.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Tubing Size: 4 mm ID
For use with Masterflex® L/S® PTFE Tubing Pump Head
This is one set consisting of two pieces.