GF Signet 155.522.554 PVDF Needle Valve, 3/8" NPT(F), with PTFE and Viton
Supplier: Avantor Fluid Handling
Create a secure pipe threaded connection.
- Constructed of durable PVDF
PVDF needle valves are pressure rated up to 200 psi (13.8 bar) with a max temperature rating of 302 deg F (150 deg C). Seats are made of PTFE and O-rings made of Viton®. Valves feature standard pipe thread connections and are autoclavable.