PVC Stopcock with Flow Indication, 1/4" NPT(M) x 1/4" NPT(M)
Supplier: Avantor Fluid Handling
Easily control flow with flow indicator handle.
- Constructed of durable PVC and EPDM
PVC bodies, EPDM seals and seats. Flow indicator on handle. Maximum pressure is 150 psi at the maximum temperature of 120 °F (49 °C); maximum vacuum is 29.9" Hg.