Masterflex® Fitting, PFA, Straight, Hose Barb to Threaded Adapter, 3/8" ID x 1/4" NPT(M)
Supplier: Avantor Fluid Handling
VWR Item#MFLX06383-93
Your Organization's Price:
Retrieving
Quick and easy assembly.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 3/8" ID x 1/4" NPT(M)
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.