Tygon Transfer Tubing, Tygon® E-Food (B-44-4X), 1/16" ID x 3/16" OD; 50 Ft
Supplier: Avantor Fluid Handling
VWR Item#MFLX95644-01
Your Organization's Price:
Retrieving
FDA compliant for food contact.
<B>Formulation:</B> Tygon® S3-B-44-4X<br /><B>Applications:</B> Food and beverage processing and dispensing, milk and dairy processing.<br /><B>Characteristics:</B> Bio-based, phthalate-free flexible. Nonwetting, odorless, tasteless, and nontoxic. Transparent.<br /><B>Certification:</B> Resin is FDA compliant, NSF-listed (Standard 51), and meets 3A standards (20-27) for food and beverage handling.<br /><B>Temperature range:</B> –49 to 165 °F (–45 to 74 °C)<br /><B>Sterilize:</B> Autoclave for 30 min at 250 °F, 15 psi or ethylene oxide.