Masterflex® Low-Density Polyethylene (LDPE) Tubing, 11/64" x 1/4"; 100 Ft
Supplier: Avantor Fluid Handling
VWR Item#MFLX95876-03
Your Organization's Price:
Retrieving
General low pressure and temperature tubing.
Applications: General low pressure and temperature, instrumentation, corrosive atmosphere, and food processing applications.<br />Characteristics: Broad chemical, solvent, and corrosive atmosphere resistance with good dimensional stability. Translucent.<br />Certification: FDA-compliant (21 CFR 177.1520); meets USDA requirements<br />Temperature range: –100 to 175 °F (–73 to 80 °C)<br />Sterilize: Ethylene oxide.