Masterflex® Drum Pump Barrel Adapter Replacement, Polypropylene
Supplier: Avantor Fluid Handling
VWR Item#MFLX70604-92
Your Organization's Price:
Retrieving
Extend the life of your drum pump!
  • Optimize performance of your Masterflex® Drum and Container Pumps
Replacement parts are compatible with Masterflex® Complete Drum and Container Pumps, 70613-series.