Masterflex® Transfer Tubing, Fluorinated Ethylene Propylene, 3/8” ID x 1/2” OD; 100 ft
Supplier: Avantor Fluid Handling
Maximize tubing on hand.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Single-use custom lengths and configurations available to minimise downtime
- Tubing size: 3/8” ID x 1/2” OD
This tubing is ideal for general laboratory use. They offer long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, are nontoxic. FDA compliant and made according to GMP. Temperature range is -51 to 165 °F (-46 to 73 °C). Can be sterilized with ethylene oxide or autoclave.