Masterflex® Medium Flow, 3/4" Sanitary Clamp Female, Open Flow, PVDF Housing, Viton O-Ring, Stainless Spring, 3/8"
Supplier: Avantor Fluid Handling
VWR Item#MFLX40257-16
Your Organization's Price:
Retrieving
Mechanical innovations reduce risk of fluid cross-over and contamination. A mating connection is made with a Medium Flow Male fitting.