Value Plastics® Fitting, Polypropylene, Straight, Female Luer to Hosebarb Adapter, 1/4" ID; 1000/PK
Supplier: Avantor Fluid Handling
VWR Item#MFLX30800-06BLK
Your Organization's Price:
Retrieving
Easily transition to a luer fitting.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: Female luer to 1/4" hose barb with 0,992" overall length