CPC® High-Flow Non-Spill Quick-Disconnect Fitting, Threaded Insert, Valved, 1/2" NPT(M), NSHD24008; 5/PK
Supplier: Avantor Fluid Handling
VWR Item#MFLX31603-72BLK
Your Organization's Price:
Retrieving
Virtually eliminate any loss of product with this leak-free design.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Non-spill design virtually eliminates spills, minimises downtime and enhances operator safety
  • Easy one-hand operation
These CPC® high-flow, non-spill quick-disconnect fittings virtually eliminate any loss of product with this leak-free design. Couplings are made of polypropylene with an EPDM O-ring for excellent chemical resistance. This non-spill series ensures minimal leaks and air inclusions in your process line - protects process purity and minimises loss due to accidental disconnects. These NSH series couplings feature a 100% springless and metal-free flow path.<br /><br />A complete coupler requires a fitting body and an insert (order separately).