Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')titanium(III)
Supplier: Thermo Fisher Scientific
|
Warning
|
40621.03
40621.06
40621.03EA
0
EUR
40621.03
40621.06
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')titanium(III)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')titanium(III)
CAS No.: 181418-64-4
Molecular Formula: C33H57O6Ti
Formula Weight: 597.70
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Purple to black
MDL No.: MFCD00145466
Formula:
C₃₃H₅₇O₆Ti Boiling Pt: 75 °C (0,1 mmHg) (subl.) Storage Temperature: Ambient |
MDL Number:
MFCD00145466 CAS Number: 181418-64-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...