Ethyl-3-methyl-3-phenylglycidate (mixture of cis and trans isomers) ≥98%
Supplier: Thermo Scientific
B23654.18
B23654.30
B23654.18EA
35.4
EUR
B23654.18
B23654.30
Ethyl-3-methyl-3-phenylglycidate (mixture of cis and trans isomers) ≥98%
Ethyl 3-methyl-3-phenylglycidate (mixture of cis and trans isomers)
Purity: 98%
CAS No.: 77-83-8
Molecular Formula: C12H14O3
Formula Weight: 206.24
Flash Point: 134°(273°F)
Refractive Index: 1.5050
Boiling Point: 272-275°
Beilstein Reference: 12299
MDL No.: MFCD00022338
Formula:
C₁₂H₁₄O₃ MW: 206,24 g/mol Boiling Pt: 272…275 °C Density: 1,044 Flash Pt: 134 °C (273 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00022338 CAS Number: 77-83-8 EINECS: 201-061-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...