2-(4-Methoxyphenyl)-4-methyl-5-thiazolecarboxylic acid ≥97%

Supplier: Thermo Scientific
Warning

H54038.MD H54038.03
H54038.MDEA 0 EUR
H54038.MD H54038.03
2-(4-Methoxyphenyl)-4-methyl-5-thiazolecarboxylic acid ≥97%
2-(4-Methoxyphenyl)-4-methyl-5-thiazolecarboxylic acid

Purity: 97% CAS No.: 54001-16-0 Molecular Formula: C12H11NO3S Formula Weight: 249.28 Melting Point: 236-241° MDL No.: MFCD06857928

Formula: C₁₂H₁₁NO₃S
MW: 249,28 g/mol
Melting Pt: 236…241 °C
Storage Temperature: Ambient
MDL Number: MFCD06857928
CAS Number: 54001-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR