2-(2-Thienyl)-4-thiazolecarboxylic acid ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-(2-Thienyl)-1,3-thiazole-4-carboxylicacid
H54347.MD
H54347.03
H54347.MDEA
0
EUR
H54347.MD
H54347.03
2-(2-Thienyl)-4-thiazolecarboxylic acid ≥97%
2-(2-Thienyl)-4-thiazolecarboxylic acid
Purity: 97%
CAS No.: 24044-07-3
Molecular Formula: C8H5NO2S2
Formula Weight: 211.25
MDL No.: MFCD01936014
Formula:
C₈H₅NO₂S₂ MW: 211,25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01936014 CAS Number: 24044-07-3 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...