4,4'-Stilbenedicarboxylic acid 96%

Supplier: Thermo Scientific
Warning

Synonyms: 4,4-Stilbenedicarboxylic acid

A11839.14 A11839.03 A11839.06
A11839.14EA 1090 EUR
A11839.14 A11839.03 A11839.06
4,4'-Stilbenedicarboxylic acid 96%
4,4'-Stilbenedicarboxylic acid

Purity: 96% CAS No.: 100-31-2 Molecular Formula: C16H12O4 Formula Weight: 268.27 Melting Point: 300° MDL No.: MFCD00013994

Formula: C₁₆H₁₂O₄
MW: 268,27 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00013994
CAS Number: 100-31-2
EINECS: 202-838-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR