Phenylphosphinic acid ≥98%
Supplier: Thermo Scientific
|
Danger
|
A17453.22
A17453.36
A17453.22EA
41.1
EUR
A17453.22
A17453.36
Phenylphosphinic acid ≥98%
Phenylphosphinic acid
Purity: 98%
CAS No.: 1779-48-2
Molecular Formula: C6H7O2P
Formula Weight: 142.10
Melting Point: 83-85°
Beilstein Reference: 2802244
UN No.: 3261
MDL No.: MFCD00002131
Formula:
C₆H₇O₂P MW: 142,092 g/mol Melting Pt: 83…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002131 CAS Number: 1779-48-2 EINECS: 217-217-3 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...