2-Amino-4-(2-bromophenyl)pyrimidine 95%

Supplier: Thermo Scientific
Warning

H34212.03 H34212.06
H34212.03EA 77.8 EUR
H34212.03 H34212.06
2-Amino-4-(2-bromophenyl)pyrimidine 95%
2-Amino-4-(2-bromophenyl)pyrimidine
Formula: C₁₀H₈BrN₃
MW: 250,10 g/mol
Melting Pt: 167…168 °C
Storage Temperature: Ambient
MDL Number: MFCD07381905
CAS Number: 99073-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR