Methyldiphenylphosphine oxide ≥98%
Supplier: Thermo Scientific
|
Warning
|
A11484.06
A11484.14
A11484.06EA
57.4
EUR
A11484.06
A11484.14
Methyldiphenylphosphine oxide ≥98%
Methyldiphenylphosphine oxide
Purity: 98+%
CAS No.: 2129-89-7
Molecular Formula: C13H13OP
Formula Weight: 216.22
Melting Point: 111-114°
Beilstein Reference: 957291
Formula:
C₁₃H₁₃OP MW: 216,22 g/mol Melting Pt: 111…114 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002081 CAS Number: 2129-89-7 EINECS: 218-346-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...