Methyl isocyanoacetate 95%
Supplier: Thermo Fisher Scientific
|
Danger
|
H31069.03
H31069.06
H31069.03EA
46
EUR
H31069.03
H31069.06
Methyl isocyanoacetate 95%
Methyl isocyanoacetate
Purity: 95%
CAS No.: 39687-95-1
Molecular Formula: C4H5NO2
Formula Weight: 99.09
Storage Temperature: 2°C to 8°C
Flash Point: 84°(183°F)
Boiling Point: 75-76°/10mm
UN No.: 2922
MDL No.: MFCD00000006
Formula:
C₄H₅NO₂ Boiling Pt: 75…76 °C (10 mmHg) Density: 1,09 Flash Pt: 84 °C (183 °F) |
MDL Number:
MFCD00000006 CAS Number: 39687-95-1 EINECS: 254-593-8 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...