N-Boc-ethylenediamine hydrochloride 98%
Supplier: Thermo Scientific
Synonyms:
tert-Butyl (2-aminoethyl)carbamate hydrochloride
L19485.03
L19485.06
L19485.03EA
85.9
EUR
L19485.03
L19485.06
N-Boc-ethylenediamine hydrochloride 98%
N-Boc-ethylenediamine hydrochloride
Purity: 98%
CAS No.: 79513-35-2
Molecular Formula: (CH3)3COCONHCH2CH2NH2.HCl
Formula Weight: 196.68
Melting Point: 148-151°
Beilstein Reference: 3627511
Formula:
C₇H₁₇ClN₂O₂ MW: 196,68 g/mol Melting Pt: 148…151 °C Storage Temperature: Ambient |
MDL Number:
MFCD01076129 CAS Number: 79513-35-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...