Potassium 2-phenylethyltrifluoroborate ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: Potassium Trifluoro(phenethyl)borate, Potassium Trifluoro(2-phenylethyl)borate, Potassium Phenethyltrifluoroborate

H25930.06 H25930.03
H25930.06EA 200 EUR
H25930.06 H25930.03
Potassium 2-phenylethyltrifluoroborate ≥98%
Potassium 2-phenylethyltrifluoroborate

Purity: 98% CAS No.: 329976-74-1 Molecular Formula: C8H9BF3K Formula Weight: 212.06 Melting Point: 173-175° MDL No.: MFCD09039257

Formula: C₈H₉BF₃K
MW: 212,06 g/mol
Melting Pt: 173…175 °C
Storage Temperature: Ambient
MDL Number: MFCD09039257
CAS Number: 329976-74-1
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR