3-Amino-3-(3-methoxyphenyl)propionic acid 97%
Supplier: Thermo Scientific
Synonyms:
3-Amino-3-(3-methoxyphenyl)propanoic acid
L19800.03
L19800.MD
L19800.03EA
97.4
EUR
L19800.03
L19800.MD
3-Amino-3-(3-methoxyphenyl)propionic acid 97%
3-Amino-3-(3-methoxyphenyl)propionic acid
Purity: 97%
CAS No.: 68208-19-5
Molecular Formula: C10H13NO3
Formula Weight: 195.22
Storage Temperature: 2°C to 8°C
Melting Point: 208° dec.
Beilstein Reference: 2838792
MDL No.: MFCD00462569
Formula:
C₁₀H₁₃NO₃ MW: 195,22 g/mol Melting Pt: 208 °C (dec.) |
MDL Number:
MFCD00462569 CAS Number: 68208-19-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...