Methyl thiosalicylate ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Methyl 2-mercaptobenzoate
H28400.14
H28400.06
H28400.14EA
257
EUR
H28400.14
H28400.06
Methyl thiosalicylate ≥97%
Methyl thiosalicylate
Purity: 97%
CAS No.: 4892-02-8
Molecular Formula: C8H8O2S
Formula Weight: 168.21
Flash Point:
Formula:
C₈H₈O₂S MW: 168,21 g/mol Boiling Pt: 98…100 °C (2 mmHg) Density: 1,223 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00060678 CAS Number: 4892-02-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...