N-Cyclohexyl-4-fluorobenzamide ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
H57640.MD
H57640.03
H57640.MDEA
0
EUR
H57640.MD
H57640.03
N-Cyclohexyl-4-fluorobenzamide ≥97%
N-Cyclohexyl 4-fluorobenzamide
Formula:
C₁₃H₁₆FNO Storage Temperature: Ambient |
MDL Number:
MFCD00544070 CAS Number: 2342-50-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...