Potassium-4-tert-butylphenyltrifluoroborate 96%

Supplier: Thermo Fisher Scientific
Danger

H52885.03 H52885.06
H52885.03EA 0 EUR
H52885.03 H52885.06
Potassium-4-tert-butylphenyltrifluoroborate 96%
Potassium 4-tert-butylphenyltrifluoroborate
Formula: C₁₀H₁₃BF₃K
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD06659940
CAS Number: 423118-47-2
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR