2-Ethoxy-5-(trifluoromethyl)phenylboronic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Ethoxy-5-(trifluoromethyl)benzeneboronic acid, 2-Ethoxy-5-trifluoromethylphenylboronic acid

H52541.MD H52541.03
H52541.MDEA 53 EUR
H52541.MD H52541.03
2-Ethoxy-5-(trifluoromethyl)phenylboronic acid ≥98%
2-Ethoxy-5-(trifluoromethyl)phenylboronic acid

Purity: 98% CAS No.: 850593-10-1 Molecular Formula: C9H10BF3O3 Formula Weight: 233.98 Melting Point: 100-104° MDL No.: MFCD06801695

Formula: C₉H₁₀BF₃O₃
MW: 233,98 g/mol
Melting Pt: 100…104 °C
Storage Temperature: Ambient
MDL Number: MFCD06801695
CAS Number: 850593-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR