2,5-Bis(chloromethyl)-4-(2-ethylhexyloxy)anisole ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
1,4-Bis(chloromethyl)-2-((2-ethylhexyl)oxy)-5-methoxybenzene
H51041.03
H51041.03EA
158
EUR
H51041.03
2,5-Bis(chloromethyl)-4-(2-ethylhexyloxy)anisole ≥98%
2,5-Bis(chloromethyl)-4-(2-ethylhexyloxy)anisole
Purity: 98%
CAS No.: 146370-52-7
Molecular Formula: C17H26Cl2O2
Formula Weight: 333.29
UN No.: 1759
MDL No.: MFCD03093939
Formula:
C₁₇H₂₆Cl₂O₂ MW: 333,29 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03093939 CAS Number: 146370-52-7 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...