Tungsten disilicide ≥99.5% (metals basis)
Supplier: Thermo Scientific
Synonyms:
Tungsten silicide
11551.09
11551.18
11551.09EA
49.2
EUR
11551.09
11551.18
Tungsten disilicide ≥99.5% (metals basis)
Tungsten disilicide
Purity: 99.5%
CAS No.: 12039-88-2
Molecular Formula: WSi2
Formula Weight: 240.02
Physical Form: -325 Mesh Powder
Odour: Odourless
MDL No.: MFCD00049704
Formula:
Si₂W MW: 240,02 g/mol Melting Pt: 2165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00049704 CAS Number: 12039-88-2 EINECS: 234-909-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...