Dinickel silicide ≥99% (metals basis excluding Co) (max. 0.1-1% Co)
Supplier: Thermo Fisher Scientific
|
Danger
|
36251.18
36251.30
36251.18EA
129
EUR
36251.18
36251.30
Dinickel silicide ≥99% (metals basis excluding Co) (max. 0.1-1% Co)
Dinickel silicide
Purity: 99%
CAS No.: 12059-14-2
Molecular Formula: Ni2Si
Formula Weight: 145.47
Physical Form: -20 Mesh Powder
Odour: Odourless
UN No.: 3077
MDL No.: MFCD00151367
Formula:
Ni₂Si Storage Temperature: Ambient |
MDL Number:
MFCD00151367 CAS Number: 12059-14-2 EINECS: 235-033-1 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...