4-Bromo-2,5-dichloro-3-thiophenesulphonyl chloride ≥97%
Supplier: Thermo Fisher Scientific
A12533.03
A12533.06
A12533.03EA
0
EUR
A12533.03
A12533.06
4-Bromo-2,5-dichloro-3-thiophenesulphonyl chloride ≥97%
4-Bromo-2,5-dichloro-3-thiophenesulphonyl chloride
Purity: 97%
CAS No.: 166964-36-9
Molecular Formula: C4BrCl3O2S2
Formula Weight: 330.44
Melting Point: 67-71°
Boiling Point: 119-121°/1mm
UN No.: 3261
MDL No.: MFCD00051664
Formula:
C₄BrCl₃O₂S₂ Melting Pt: 67…71 °C |
MDL Number:
MFCD00051664 CAS Number: 166964-36-9 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...