2-Chloro-3-nitro-5-thiophenesulphonyl chloride ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
5-Chloro-4-nitrothiophene-2-sulphonyl chloride
A11779.06
A11779.03
A11779.06EA
148
EUR
A11779.06
A11779.03
2-Chloro-3-nitro-5-thiophenesulphonyl chloride ≥98%
2-Chloro-3-nitro-5-thiophenesulphonyl chloride
Purity: 98%
CAS No.: 58457-24-2
Molecular Formula: C4HCl2NO4S2
Formula Weight: 262.09
Melting Point: 52-54°
UN No.: 3261
MDL No.: MFCD00067978
Formula:
C₄HCl₂NO₄S₂ MW: 262,09 g/mol Melting Pt: 52…54 °C Storage Temperature: Ambient |
MDL Number:
MFCD00067978 CAS Number: 58457-24-2 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...