Thiophene-2-methylamine 97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-Thiophenemethylamine, 2-(Aminomethyl)thiophene
A11700.06
A11700.14
A11700.22
A11700.06EA
50.1
EUR
A11700.06
A11700.14
A11700.22
Thiophene-2-methylamine 97%
Thiophene-2-methylamine
Purity: 97%
CAS No.: 27757-85-3
Molecular Formula: C5H7NS
Formula Weight: 113.19
Flash Point: 73°(163°F)
Refractive Index: 1.5670
Boiling Point: 85-88°/14mm
Beilstein Reference: 106296
UN No.: 2735
MDL No.: MFCD00005460
Formula:
C₅H₇NS MW: 113,19 g/mol Boiling Pt: 85…88 °C (14 mmHg) Density: 1,100 Flash Pt: 73 °C (163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005460 CAS Number: 27757-85-3 EINECS: 248-639-6 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...