2,1,3-Benzothiadiazole-5-carboxylic acid ≥97%

Supplier: Thermo Scientific
Warning

H33095.MD H33095.03 H33095.06
H33095.MDEA 70 EUR
H33095.MD H33095.03 H33095.06
2,1,3-Benzothiadiazole-5-carboxylic acid ≥97%
2,1,3-Benzothiadiazole-5-carboxylic acid
Formula: C₇H₄N₂O₂S
MW: 180,19 g/mol
Melting Pt: 228…230 °C
Storage Temperature: Ambient
MDL Number: MFCD01647555
CAS Number: 16405-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR