Ethyl-3-ethoxypropionate 99+% stabilised, pure
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Ethyl 3-ethoxypropanoate
269490010
269490025
269490100
ACRO269490010EA
61.4
EUR
ACRO269490010
ACRO269490025
ACRO269490100
Ethyl-3-ethoxypropionate 99+% stabilised, pure
Ethyl 3-ethoxypropionate
Formula:
C₇H₁₄O₃ MW: 146,19 g/mol Boiling Pt: 170 °C (760 mmHg) Melting Pt: –75 °C Density: 0,949 g/cm³ Flash Pt: 59 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051356 CAS Number: 763-69-9 EINECS: 212-112-9 UN: 3272 ADR: 3,III |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4060 to 1.4080 (20°C, 589 nm) |
Water | ≤0.05 % |
GC | ≥99.0 % |
Color scale | ≤15 APHA |
Stabilizer | 50 to 100 ppm BHT |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...