Ethyl-3-ethoxypropionate ≥99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Ethyl 3-ethoxypropanoate
B24024.AK
B24024.AU
B24024.AKEA
30.6
EUR
B24024.AK
B24024.AU
Ethyl-3-ethoxypropionate ≥99%
Ethyl 3-ethoxypropionate
Purity: 99+%
CAS No.: 763-69-9
Molecular Formula: C7H14O3
Formula Weight: 146.19
Flash Point: 59°(142°F)
Refractive Index: 1.4065
Odour: Ester-like
Melting Point: -100°
Boiling Point: 169-171°
UN No.: 3272
MDL No.: MFCD00051356
Formula:
C₇H₁₄O₃ MW: 146,19 g/mol Boiling Pt: 169…171 °C Melting Pt: –100 °C Density: 0,949 Flash Pt: 59 °C (142 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051356 CAS Number: 763-69-9 EINECS: 212-112-9 UN: 3272 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...