4-Bromo-3-(trifluoromethyl)anisole ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
H31682.03
H31682.09
H31682.03EA
0
EUR
H31682.03
H31682.09
4-Bromo-3-(trifluoromethyl)anisole ≥98%
4-Bromo-3-(trifluoromethyl)anisole
Purity: 98%
CAS No.: 400-72-6
Molecular Formula: C8H6BrF3O
Formula Weight: 255.04
MDL No.: MFCD08059527
Formula:
C₈H₆BrF₃O Storage Temperature: Ambient |
MDL Number:
MFCD08059527 CAS Number: 400-72-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...