3-(2,4,6-Trimethoxyphenyl)propionic acid 96%
Supplier: Thermo Fisher Scientific
|
Warning
|
H34208.03
H34208.09
H34208.03EA
0
EUR
H34208.03
H34208.09
3-(2,4,6-Trimethoxyphenyl)propionic acid 96%
3-(2,4,6-Trimethoxyphenyl)propionic acid
Purity: 96%
CAS No.: 74737-05-6
Molecular Formula: C12H16O5
Formula Weight: 240.26
Melting Point: 140-144°
MDL No.: MFCD00045944
Formula:
C₁₂H₁₆O₅ Melting Pt: 140…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD00045944 CAS Number: 74737-05-6 EINECS: 277-978-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...