4-Methyl-1H-pyrazole ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
A18083.03
A18083.06
A18083.03EA
57.3
EUR
A18083.03
A18083.06
4-Methyl-1H-pyrazole ≥97%
4-Methyl-1H-pyrazole
Purity: 97+%
CAS No.: 7554-65-6
Molecular Formula: C4H6N2
Formula Weight: 82.11
Flash Point: 96°(204°F)
Refractive Index: 1.4945
Melting Point: 13°
Boiling Point: 204-207°
Beilstein Reference: 105204
Merck Reference: 14,4216
MDL No.: MFCD00005245
Formula:
C₄H₆N₂ Boiling Pt: 204…207 °C Melting Pt: 13 °C Density: 1,018 Flash Pt: 96 °C (204 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005245 CAS Number: 7554-65-6 EINECS: 231-445-0 Merck Index: 14,4216 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...