4-Bromo-2-(trifluoromethyl)benzoic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

H31656.03 H31656.06
H31656.03EA 92.4 EUR
H31656.03 H31656.06
4-Bromo-2-(trifluoromethyl)benzoic acid ≥98%
4-Bromo-2-(trifluoromethyl)benzoic acid

Purity: 98% CAS No.: 320-31-0 Molecular Formula: C8H4BrF3O2 Formula Weight: 269.02 MDL No.: MFCD07787468

Formula: C₈H₄BrF₃O₂
MW: 269.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07787468
CAS Number: 320-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR